Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylselenyl chloride, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
25g
50g
Packaging:
Glass bottle
5707-04-0
C6H5ClSe
191.52
MFCD00000478
WJCXADMLESSGRI-UHFFFAOYSA-N
phenylselenenyl chloride,phenylselenyl chloride,benzeneselenenyl chloride,chloroselenobenzene,phenyl hypochloroselenoite,phenylselenium chloride,phsecl,phenylselenenylchloride,phenylselanyl chlorane,chloroselanylbenzene
21928
phenyl selenohypochlorite
C1=CC=C(C=C1)[Se]Cl
5707-04-0
C6H5ClSe
191.52
MFCD00000478
WJCXADMLESSGRI-UHFFFAOYSA-N
phenylselenenyl chloride,phenylselenyl chloride,benzeneselenenyl chloride,chloroselenobenzene,phenyl hypochloroselenoite,phenylselenium chloride,phsecl,phenylselenenylchloride,phenylselanyl chlorane,chloroselanylbenzene
21928
phenyl selenohypochlorite
C1=CC=C(C=C1)[Se]Cl
C-X bond formation reagent
Melting Point 61.0°C to 65.0°C
Color Orange
Quantity 50g
Infrared Spectrum Authentic
Boiling Point 120.0°C (20.0 mmHg)
Packaging Glass bottle
Assay Percent Range 97.5% min. (Argentometry)
Linear Formula C6H5SeCl
Beilstein 06, III, 1110
Fieser 05,518; 06,459; 07,286; 09,25; 11,33; 13,26; 16,19; 17,26
Formula Weight 191.52
Physical Form Crystals or Crystalline Powder
Percent Purity 98%
Chemical Name or Material Phenylselenyl chloride, 98%
Show More Show Less

GHS H Statement
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Danger

EINECSNumber : 227-196-2