Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Phenylsilane, 97%

Click to view available options
Quantity:
5g
25g
100g
694-53-1
C6H5Si
105.191
MFCD00013478
XJWOWXZSFTXJEX-UHFFFAOYSA-N
phenylsilane,benzene, silyl,silylbenzene,silane, phenyl,fenylsilan,fenylsilan czech,phenylsilane, silyl,silane, phenyl-,
6327628
phenylsilicon
C1=CC=C(C=C1)[Si]
694-53-1
C6H5Si
105.191
MFCD00013478
XJWOWXZSFTXJEX-UHFFFAOYSA-N
phenylsilane,benzene, silyl,silylbenzene,silane, phenyl,fenylsilan,fenylsilan czech,phenylsilane, silyl,silane, phenyl-,
6327628
phenylsilicon
C1=CC=C(C=C1)[Si]
Applications
It is used as a pharmaceutical intermediate.

Solubility
Contact with water releases a flammable alcohol. Soluble in water (Contact with water releases flammable gases), and most common organic solvents.

Notes
Moisture Sensitive. Store away from water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect against electrostatic charges. Keep ignition sources away. Store under inert gas.
UN Number UN3399
Solubility Information Contact with water releases a flammable alcohol. Soluble in water (Contact with water releases flammable gases),and most common organic solvents.
Formula Weight 108.22
Percent Purity 97%
Density 0.88
Refractive Index 1.5105
Sensitivity Moisture sensitive
Boiling Point 120°C
Quantity 100g
Flash Point 7°C (45°F)
Chemical Name or Material Phenylsilane
Show More Show Less

GHS H Statement
H225-H261-H302-H332-H315-H319-H335
Highly flammable liquid and vapour.
In contact with water releases flammable gas.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P335+P334-P363-P370+P378q-P50

H225-H261-H302+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE

EINECSNumber : 211-772-5

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen