Alfa Aesar™ Phenylsuccinic anhydride, 99%

Manufacturer: Alfa Aesar™ B2441814

Quantity 25g

 View more versions of this product

Catalog No. AAB2441814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1131-15-3
    Molecular Formula C10H8O3
    Formula Weight 176.17
    MDL Number MFCD00192395
    Chemical Name or Material Phenylsuccinic anhydride

    Specifications

    Formula Weight 176.17
    Molecular Formula C10H8O3
    MDL Number MFCD00192395
    Boiling Point 191° to 192°C (12mmHg)
    Melting Point 53° to 55°C
    Quantity 25g
    CAS 1131-15-3
    Chemical Name or Material Phenylsuccinic anhydride
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit