Alfa Aesar™ Phenyltin trichloride

Manufacturer: Alfa Aesar™ 7115714

Quantity 25g

 View more versions of this product

Catalog No. AA7115714

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1124-19-2
    Molecular Formula C6H5SnCl3
    Formula Weight 302.18
    MDL Number MFCD00000513
    UN Number UN2788
    Synonym Trichlorophenylstannane; Trichlorophenyltin
    Chemical Name or Material Phenyltin trichloride

    Specifications

    Formula Weight 302.18
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    Synonym Trichlorophenylstannane; Trichlorophenyltin
    Health Hazard 2 GHS H Statement
    H301-H311-H331-H314-H318
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Odor Pungent
    MDL Number MFCD00000513
    CAS 1124-19-2
    Quantity 25g
    UN Number UN2788
    Health Hazard 1 Danger
    Density 1.84g/mL
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
    Sensitivity Moisture Sensitive
    Molecular Formula C6H5SnCl3
    Chemical Name or Material Phenyltin trichloride

    GHS H Statement
    H301-H311-H331-H314-H318
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit