Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phenyltin Trichloride 98.0+%, TCI America™
Click to view available options
Quantity:
5 g
25 g
Specifications
Color | Yellow |
Boiling Point | 143°C |
UN Number | 3265 |
Quantity | 25 g |
Formula Weight | 302.17 |
Percent Purity | ≥98.0% (T) |
Physical Form | Liquid |
Chemical Name or Material | Phenyltin Trichloride |
Safety and Handling
RTECSNumber : WH8730000
TSCA : No