Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyltin Trichloride 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
1124-19-2
C6H5Cl3Sn
302.166
MFCD00000513
UBOGEXSQACVGEC-UHFFFAOYSA-K
phenyltin trichloride, phenyltrichlorotin, stannane, trichlorophenyl, trichlorophenyltin, trichloro phenyl stannane, phenyltrichlorostannane, ccris 6323, phsncl3, phenyl tin trichloride, monophenyltin trichloride
14298
trichloro(phenyl)stannane
C1=CC=C(C=C1)[Sn](Cl)(Cl)Cl
1124-19-2
C6H5Cl3Sn
302.166
MFCD00000513
UBOGEXSQACVGEC-UHFFFAOYSA-K
phenyltin trichloride, phenyltrichlorotin, stannane, trichlorophenyl, trichlorophenyltin, trichloro phenyl stannane, phenyltrichlorostannane, ccris 6323, phsncl3, phenyl tin trichloride, monophenyltin trichloride
14298
trichloro(phenyl)stannane
C1=CC=C(C=C1)[Sn](Cl)(Cl)Cl
Color Yellow
Boiling Point 143°C
UN Number 3265
Quantity 5g
Formula Weight 302.17
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (T)
Chemical Name or Material Phenyltin Trichloride

RTECSNumber : WH8730000

TSCA : No