Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyltrichlorosilane 95%, ACROS Organics™

Click to view available options
Quantity:
10mL
250mL
1L
Packaging:
Glass bottle
98-13-5
C6H5Cl3Si
211.55
MFCD00000479
ORVMIVQULIKXCP-UHFFFAOYSA-N
trichloro phenyl silane,phenyltrichlorosilane,silane, trichlorophenyl,phenyl trichlorosilane,phenylsilicon trichloride,silane, phenyltrichloro,silicon phenyl trichloride,benzene, trichlorosilyl,unii-m7199jl06m,phenyl-trichloro-silane
7372
trichloro(phenyl)silane
C1=CC=C(C=C1)[Si](Cl)(Cl)Cl
98-13-5
C6H5Cl3Si
211.55
MFCD00000479
ORVMIVQULIKXCP-UHFFFAOYSA-N
trichloro phenyl silane,phenyltrichlorosilane,silane, trichlorophenyl,phenyl trichlorosilane,phenylsilicon trichloride,silane, phenyltrichloro,silicon phenyl trichloride,benzene, trichlorosilyl,unii-m7199jl06m,phenyl-trichloro-silane
7372
trichloro(phenyl)silane
C1=CC=C(C=C1)[Si](Cl)(Cl)Cl
Refractive Index 1.5220 to 1.524
Boiling Point 201°C
Flash Point 91°C
Specific Gravity 1.321
Packaging Glass bottle
Color Colorless to Yellow
Density 1.3210g/mL
Infrared Spectrum Authentic
Quantity 250mL
Assay Percent Range 95%
Linear Formula C6H5SiCl3
Beilstein 16, 911
Solubility Information Solubility in water: reacts. Other solubilities: soluble in benzene,ehter and perchloroethylene
Formula Weight 211.55
Physical Form Liquid
Percent Purity ≥94%
Chemical Name or Material Phenyltrichlorosilane, 95%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Fatal if inhaled.
Reacts violently with water.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair).

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 202-640-8

RTECSNumber : VV6650000

TSCA : TSCA