Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyltriethoxysilane, 98%, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
Packaging:
Glass bottle
780-69-8
C12H20O3Si
240.38
MFCD00009065
JCVQKRGIASEUKR-UHFFFAOYSA-N
phenyltriethoxysilane,triethoxy phenyl silane,silane, triethoxyphenyl,triethoxyfenylsilan,silane, phenyltriethoxy,benzeneorthosiliconic acid, triethyl ester,phenyl triethoxysilane,benzene, triethoxysilyl,triethoxyfenylsilan czech,unii-qi310t2x15
13075
triethoxy(phenyl)silane
CCO[Si](C1=CC=CC=C1)(OCC)OCC
780-69-8
C12H20O3Si
240.38
MFCD00009065
JCVQKRGIASEUKR-UHFFFAOYSA-N
phenyltriethoxysilane,triethoxy phenyl silane,silane, triethoxyphenyl,triethoxyfenylsilan,silane, phenyltriethoxy,benzeneorthosiliconic acid, triethyl ester,phenyl triethoxysilane,benzene, triethoxysilyl,triethoxyfenylsilan czech,unii-qi310t2x15
13075
triethoxy(phenyl)silane
CCO[Si](C1=CC=CC=C1)(OCC)OCC
Packaging Glass bottle
Flash Point 42°C
Infrared Spectrum Authentic
Density 0.9960g/mL
Boiling Point 235.0°C
Refractive Index 1.4594 to 1.4614
Quantity 250g
Specific Gravity 0.996
Color Colorless
Assay Percent Range 98%
Linear Formula C6H5Si(OC2H5)3
Beilstein 16, 911
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 240.38
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Phenyltriethoxysilane, 98%
Show More Show Less

GHS H Statement
Flammable liquid and vapor.
May cause damage to organs through prolonged or repeated exposure if inhaled.

GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.

GHS Signal Word: Warning

EINECSNumber : 212-305-8

RTECSNumber : VV4900000

TSCA : TSCA