Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyltrimethylammonium chloride, 98+%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass Bottle
138-24-9
C9H14ClN
171.668
MFCD00011790
MQAYPFVXSPHGJM-UHFFFAOYSA-M
phenyltrimethylammonium chloride,n,n,n-trimethylbenzenaminium chloride,trimethylphenylammonium chloride,n,n,n-trimethylanilinium chloride,ammonyx 200,benzenaminium, n,n,n-trimethyl-, chloride,trimethylphenylammoniumchloride,trimethylanilinium chloride,ammonium, phenyltrimethyl-, chloride,trimethylphenyl ammonium chloride
67309
trimethyl(phenyl)azanium;chloride
C[N+](C)(C)C1=CC=CC=C1.[Cl-]
138-24-9
C9H14ClN
171.668
MFCD00011790
MQAYPFVXSPHGJM-UHFFFAOYSA-M
phenyltrimethylammonium chloride,n,n,n-trimethylbenzenaminium chloride,trimethylphenylammonium chloride,n,n,n-trimethylanilinium chloride,ammonyx 200,benzenaminium, n,n,n-trimethyl-, chloride,trimethylphenylammoniumchloride,trimethylanilinium chloride,ammonium, phenyltrimethyl-, chloride,trimethylphenyl ammonium chloride
67309
trimethyl(phenyl)azanium;chloride
C[N+](C)(C)C1=CC=CC=C1.[Cl-]
Phase transfer catalyst
Infrared Spectrum Authentic
Quantity 25g
Color Blue-Gray or White
Packaging Glass Bottle
Assay Percent Range 98+%
Linear Formula C6H5N(CH3)3Cl
Beilstein 12, 158
Formula Weight 171.67
Physical Form Crystalline Powder
Percent Purity ≥98.0%
Chemical Name or Material Phenyltrimethylammonium chloride, 98+%
Show More Show Less

GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 205-319-0

RTECSNumber : BT2190000

TSCA : TSCA