Learn More
Aviva Systems Biology Corp PHF10 Antibody - middle region (ARP48833_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP48833_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PHF10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PHF10. Alternate names: FLJ10975, MGC111009, XAP135, BAF45AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59932