Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHF11 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP18005420UL

 View more versions of this product

Catalog No. NBP18005420


Add to cart

Description

Description

PHF11 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Immunohistochemistry, Western Blotting.
Specifications

Specifications

PHF11
Unconjugated
PBS & 2% Sucrose. with No Preservative
Affinity Purified
PHF11
Synthetic peptide directed towards the middle region of human PHF11. Peptide sequence FSGVKRKRGRKKPLSGNHVQPPETMKCNTFIRQVKEEHGRHTDATVKVPF.
37 kDa
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Immunohistochemistry, Western Blot
Western Blot 1:1000, Immunohistochemistry
NP_001035533
APY, BCAPIGHER, BRCA1 C-terminus-associated protein, IgE responsiveness (atopic), IGEL, IGER, NYREN34, NY-REN-34, NY-REN-34 antigen, PHD finger protein 11, Renal carcinoma antigen NY-REN-34
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
51131
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit