Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PHF12 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548127
Description
Predicted to react with bovine, chicken, pig and rat based on sequence homology.
Acts as a transcriptional repressor. Involved in recruitment of functional SIN3A complexes to DNA. Represses transcription at least in part through the activity of an associated histone deacetylase (HDAC). May also repress transcription in a SIN3A-independent manner through recruitment of functional AES complexes to DNA.Specifications
PHF12 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q96QT6, Q5SPL2 | |
PHF12 | |
KLH conjugated synthetic peptide between 831-859 amino acids from the C-terminal region of human PHF12. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
PHF12 | |
PHD finger protein 12, PHD factor 1, Pf1, PHF12 (HGNC:20816) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
268448, 57649 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction