Learn More
PHF17 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 150321AP
Catalog No. 50-172-9919
Add to cart
Description
Description
Component of the HBO1 complex which has a histone H4-specific acetyltransferase activity, a reduced activity toward histone H3 and is responsible for the bulk of histone H4 acetylation in vivo. Transcriptional coactivator, it may also promote acetylation of nucleosomal histone H4 by KAT5. Promotes apoptosis. May act as a renal tumor suppressor. Negatively regulates canonical Wnt signaling; at least in part, cooperates with NPHP4 in this function.
Specifications
Specifications
PHF17
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6IE81, Q6ZPI0
JADE1, KIAA1807, PHD finger protein 17, PHF17, Protein Jade 1
Rabbit
IgG
150 μL
-20°C
Primary
269424, 310352, 79960
Western Blot
Unconjugated
JADE1
Liquid
Jade1
PHF17 Fusion Protein Ag4290
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat