Learn More
Aviva Systems Biology Corp PHF19 Antibody - C-terminal region (ARP50354_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP50354_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PHF19. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PHF19. Alternate names: MGC131698, MGC149712, MGC149713, MGC23929, PCL3, MTF2L1ApplicationWB. Species reactivity: Human, Yeast, Zebrafish
Catalog No. 50-014-60324