Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PHF2 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB300162FR
Description
PHF2 Polyclonal antibody specifically detects PHF2 in Human samples. It is validated for Western BlotSpecifications
PHF2 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 680 | |
GRC5, jumonji C domain-containing histone demethylase 1E, KIAA0662JHDM1E, MGC176680, PHD finger protein 2 | |
Internal peptide residues made to the 120 kDa PHF protein. | |
0.1 mL | |
Neuroscience | |
5253 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction