Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHF2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$177.16 - $378.00

Specifications

Antigen PHF2
Concentration 1 mg/mL
Dilution Western Blot 1:500 -1:1000
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB300162
View Documents
Novus Biologicals
NB300162
0.1 mg
Each for $378.00
Add to cart
 
NB018143
View Documents
Novus Biologicals
NB300162SS
0.025 mg
Each for $177.16
Add to cart
 
Description

Description

PHF2 Polyclonal specifically detects PHF2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

PHF2
Western Blot 1:500 -1:1000
Polyclonal
Rabbit
Neuroscience
PBS with 0.02% Sodium Azide
5253
Internal peptide residues made to the 120 kDa PHF protein.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
1 mg/mL
Western Blot
Unconjugated
RUO
Human
GRC5, jumonji C domain-containing histone demethylase 1E, KIAA0662JHDM1E, MGC176680, PHD finger protein 2
PHF2
IgG
Affinity Purified
Reacts with residues 70-82 [KHGPGPTPDVKRVC] of the 120 kDa PHF protein.
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit