Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PHF2 Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Specifications
Antigen | PHF2 |
---|---|
Applications | Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB029197
|
Novus Biologicals
NBP293667100UL |
100 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB029198
|
Novus Biologicals
NBP29366720UL |
20 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
PHF2 Polyclonal antibody specifically detects PHF2 in Human, Rat samples. It is validated for ImmunofluorescenceSpecifications
PHF2 | |
Polyclonal | |
Purified | |
RUO | |
Human, Rat | |
GRC5, jumonji C domain-containing histone demethylase 1E, KIAA0662JHDM1E, MGC176680, PHD finger protein 2 | |
A synthetic peptide corresponding to a sequence within amino acids 850-950 of human PHF2 (NP_005383.3). | |
Primary | |
Store at -20°:C. Avoid freeze-thaw cycles. |
Immunofluorescence | |
Unconjugated | |
Rabbit | |
Neuroscience | |
PBS (pH 7.3), 50% glycerol | |
5253 | |
IgG | |
Affinity purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title