Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHF20L1 Mouse anti-Human, Clone: OTI6G6, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527205

Catalog No. PIMA527205


Add to cart

Description

Description

Gene ontology (GO) annotation includes regulation of transcription, DNA-templated.
Specifications

Specifications

PHF20L1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
A8MW92
PHF20L1, CGI-72, TDRD20B, URLC1
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51105
Immunohistochemistry (Paraffin), Western Blot
OTI6G6
Unconjugated
PHF20L1
Liquid
PHF20L1
Full length human recombit protein of PHF20L1 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit