Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHGDH Mouse anti-Human, Clone: OTI9C2, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526820

Catalog No. PIMA526820


Add to cart

Description

Description

3-Phosphoglycerate dehydrogenase (PHGDH; EC 1.1.1.95) catalyzes the transition of 3-phosphoglycerate into 3-phosphohydroxypyruvate, which is the first and rate-limiting step in the phosphorylated pathway of serine biosynthesis, using NAD+/NADH as a cofactor.
Specifications

Specifications

PHGDH
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
O43175
3-PGDH, 3PGDH, PDG, PGAD, PGD, PGDH, SERA, 3-phosphoglycerate dehydrogenase, D-3-phosphoglycerate dehydrogenase
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
26227
Immunohistochemistry (Paraffin), Western Blot
OTI9C2
Unconjugated
PHGDH
Liquid
PHGDH
Full length human recombit protein of PHGDH produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit