Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Phi6 RNA Replicase

Catalyzes the synthesis of a full-length complementary RNA strand using single-stranded RNA or denatured double-stranded DNA as templates.

Specifications

For Use With (Application) Replication of ssRNA to dsRNA form, synthesis and amplification of dsRNA using dsDNA as template
Includes Phi6 RNA Replicase is supplied with 10x Phi6 RNA Replicase Buffer and 50 mM MnCl2 solution.
Product Type Phi6 RNA Replicase
Storage Requirements Recommended storage temperature -20°C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
F611S
View Documents
Thermo Scientific™
F611S
60 (1U/µL) This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
F611L
View Documents
Thermo Scientific™
F611L
300 (1U/µL) This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Phi6 RNA Replicase is a modified version of the protein P2 from bacteriophage Phi6. This novel RNA-dependent RNA polymerase catalyzes the synthesis of a full-length complementary RNA strand initiating from the 3' terminus of a single-stranded RNA. The polymerase does not require any oligonucleotide primer for the initiation. Due to a unique modification, the Phi6 RNA Replicase displays relatively low template specificity, and it is therefore capable of replicating a great variety of RNA templates as well as denatured DNA that contains a recognition sequence for the Phi6 RNA Replicase at its 3' terminus.

Applications

  • Replication of ssRNA to dsRNA form
  • ssDNA replication to RNA/DNA hybrid
  • For producing larger amounts of dsRNA, Replicator RNAi Kit is recommended

Includes

  • Phi6 RNA Replicase is supplied with 10X Phi6 RNA Replicase Buffer and 50 mM MnCl2 solution. 1X buffer contains: 50 mM Tris-HCl (pH 8.75 at 21°C), 50 mM NH4Ac.
Specifications

Specifications

Replication of ssRNA to dsRNA form, synthesis and amplification of dsRNA using dsDNA as template
Phi6 RNA Replicase is supplied with 10x Phi6 RNA Replicase Buffer and 50 mM MnCl2 solution.
Phi6 RNA Replicase
Recommended storage temperature -20°C
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit