Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Phire Green Hot Start II DNA Polymerase

A novel hot start DNA Polymerase for routine and high throughput PCR applications requiring speed and specifity.

$243.00 - $1008.00

Specifications

GC-Rich PCR Performance Medium
Polymerase Phire Hot Start II DNA polymerase
Reaction Speed Fast
Product Line Phire
Product Type Hot Start High-Fidelity DNA Polymerase
View More Specs

Products 2
Catalog Number Mfr. No. Sufficient For Price Quantity & Availability  
Catalog Number Mfr. No. Sufficient For Price Quantity & Availability  
F124L
View Documents
Thermo Scientific™
F124L
1000 x 50 μL Reactions
Each for $1,008.00
Add to Cart
 
F124S
View Documents
Thermo Scientific™
F124S
200 x 50 μL Reactions
Each for $243.00
Add to Cart
 
Description

Description

Phire Hot Start II DNA Polymerase is significantly faster, extremely robust, and also capable of amplifying long DNA fragments with high yields. These features are achieved through advanced protein engineering of the polymerase. It incorporates a unique double-stranded DNA binding domain which allows short extension times (10 to 15 s/kb), improves yields, and increases fidelity 2-fold compared to Taq DNA polymerase.

The hot start modification of Phire Hot Start II DNA Polymerase is based on the same Affibody inactivation technology utilized by Phusion Hot Start II DNA Polymerase. This technology increases the specificity of the PCR reaction with no additional time required for initial activation of the enzyme.

New Phire Green format is a combination of Phire Hot Start II DNA Polymerase and 5X Phire Green Buffer. The buffer includes a density reagent and tracking dyes for direct loading of PCR products on a gel. The colored buffer does not interfere with enzyme performance and is compatible with downstream applications such as DNA sequencing, ligation and restriction digestion.

Thermo Scientific Phire Hot Start II DNA Polymerase is a novel PCR enzyme for routine and high throughput PCR applications. It outperforms every Taq-based hot start polymerase on the market. Phire Hot Start II DNA Polymerase is significantly faster, extremely robust, and also capable of amplifying long DNA fragments with high yields. These features are achieved through advanced protein engineering of the polymerase. It incorporates a unique double-stranded DNA binding domain which allows short extension times (10 to 15 s/kb), improves yields, and increases fidelity 2-fold compared to Taq DNA polymerase.
The hot start modification of Phire Hot Start II DNA Polymerase is based on the same Affibody inactivation technology utilized by Phusion Hot Start II DNA Polymerase. This technology increases the specificity of the PCR reaction with no additional time required for initial activation of the enzyme.
New Phire Green format is a combination of Phire Hot Start II DNA Polymerase and 5X Phire Green Buffer. The buffer includes a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffer does not interfere with enzyme performance and is compatible with downstream applications such as DNA sequencing, ligation and restriction digestion.


Highlights

  • Quick hot start - No reactivation step
  • Fast enzyme - Amplify four times faster than with hot start Taq
  • Robust - Minimal reaction optimization due to high inhibitor tolerance
  • High yields - Abundant products due to high efficiency
  • Longer PCR products - Amplify significantly longer DNA fragments than with any hot start Taq
  • Direct loading on gel with Green Buffer
Applications
  • Hot Start PCR
  • Routine PCR
  • Non-high fidelity PCR
  • Fast PCR
  • High throughput PCR
  • Genotyping
Includes
Phire Hot Start II DNA Polymerase
  • Phire Hot Start II DNA Polymerase
  • 5X Phire Reaction Buffer (provides 1.5 mM MgCl2 in the final 1X concentration)
  • DMSO
Phire Green Hot Start II DNA Polymerase
  • Phire Hot Start II DNA Polymerase
  • 5X Phire Green Reaction Buffer (provides 1.5 mM MgCl2 in the final 1X concentration)
  • DMSO

Specifications

Specifications

Medium
Fast
Hot Start High-Fidelity DNA Polymerase
50X
Built-In Hot Start
Standalone
Phire Hot Start II DNA polymerase
Phire
Cloning
2 X
Blunt
DNA
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit