Learn More
Thermo Scientific™ Phire™ Hot Start DNA Polymerase
Thermo Scientific™ Phire Hot Start DNA Polymerase is the older version of Phire Hot Start DNA Polymerase; for upgraded version, please see Phire Hot Start II DNA Polymerase.
Manufacturer:  Thermo Scientific™ F120S
 View more versions of this product
Catalog No. F120S
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Thermo Scientific™ Phire™ Hot Start DNA Polymerase is the older version of this polymerase. For an upgraded version, please see Phire Hot Start II DNA Polymerase.
Note: Phire Hot-Start DNA Polymerase has been upgraded to new Phire Hot Start II DNA Polymerase (F-122S and F-122L). Current Phire Hot Start customers can switch to Phire Hot Start II with no changes needed to existing protocols or primers. Original Phire Hot Start will remain available for purchase, however, all new customers are advised to purchase Phire Hot Start II to allow more flexibility in primer design.
  • Quick hot start: No reactivation step
  • Fast enzyme: Amplify four times faster than with hot start Taq
  • Robust: Minimal reaction optimization due to high inhibitor tolerance
  • High yields: Abundant products due to high efficiency
  • Longer PCR products: Amplify significantly longer DNA fragments than with any hot start Taq
Includes:
Phire Hot Start DNA Polymerase is supplied with 5x Phire Reaction Buffer and DMSO. The buffer provides 1.5 mM MgCl2 in the final 1x concentration.
Recommended for:
Fast PCR, routine PCR, high-throughput PCR, long-range PCR, DHPLC, microarray
Specifications
Specifications
Phire™ Hot Start DNA Polymerase
Phire Hot Start DNA Polymerase is supplied with 5x Phire Reaction Buffer and DMSO. The buffer provides 1.5 mM MgCl2 in the final 1x concentration.
Recommended storage temperature -20°C. Stable for one year from the assay date.
Fast PCR, routine PCR, high-throughput PCR, long-range PCR, DHPLC, microarray
200 (50µL vol) reactions