Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Phire™ Hot Start DNA Polymerase

Thermo Scientific™ Phire Hot Start DNA Polymerase is the older version of Phire Hot Start DNA Polymerase; for upgraded version, please see Phire Hot Start II DNA Polymerase.

Specifications

Description Phire™ Hot Start DNA Polymerase
For Use With (Application) Fast PCR, routine PCR, high-throughput PCR, long-range PCR, DHPLC, microarray
Includes Phire Hot Start DNA Polymerase is supplied with 5x Phire Reaction Buffer and DMSO. The buffer provides 1.5 mM MgCl2 in the final 1x concentration.
Storage Requirements Recommended storage temperature -20°C. Stable for one year from the assay date.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
F120S
View Documents
Thermo Scientific™
F120S
200 (50µL vol) reactions This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
F120L
View Documents
Thermo Scientific™
F120L
1000 (50µL vol) reactions This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Thermo Scientific™ Phire™ Hot Start DNA Polymerase is the older version of this polymerase. For an upgraded version, please see Phire Hot Start II DNA Polymerase.

Note: Phire Hot-Start DNA Polymerase has been upgraded to new Phire Hot Start II DNA Polymerase (F-122S and F-122L). Current Phire Hot Start customers can switch to Phire Hot Start II with no changes needed to existing protocols or primers. Original Phire Hot Start will remain available for purchase, however, all new customers are advised to purchase Phire Hot Start II to allow more flexibility in primer design.
  • Quick hot start: No reactivation step
  • Fast enzyme: Amplify four times faster than with hot start Taq
  • Robust: Minimal reaction optimization due to high inhibitor tolerance
  • High yields: Abundant products due to high efficiency
  • Longer PCR products: Amplify significantly longer DNA fragments than with any hot start Taq

Includes:

Phire Hot Start DNA Polymerase is supplied with 5x Phire Reaction Buffer and DMSO. The buffer provides 1.5 mM MgCl2 in the final 1x concentration.

Recommended for:

Fast PCR, routine PCR, high-throughput PCR, long-range PCR, DHPLC, microarray

Specifications

Specifications

Phire™ Hot Start DNA Polymerase
Fast PCR, routine PCR, high-throughput PCR, long-range PCR, DHPLC, microarray
Phire Hot Start DNA Polymerase is supplied with 5x Phire Reaction Buffer and DMSO. The buffer provides 1.5 mM MgCl2 in the final 1x concentration.
Recommended storage temperature -20°C. Stable for one year from the assay date.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit