Learn More
Thermo Scientific™ Phire Plant Direct PCR Master Mix Promotion
Amplify DNA directly from plant samples without prior DNA purification with Phire Plant Direct PCR Master Mix
Manufacturer:  Thermo Scientific™ F160L
 View more versions of this product
Catalog No. F160LPM
Add to cart
Description
Description
  • Direct PCR — sample is added directly to PCR reaction, therefore there is no need for time-consuming and expensive DNA purification steps.
  • Dilution ampersand (and) Storage protocol available — allows tens of PCR reactions from one tiny sample and allows re-testing
  • Specially engineered Phire Hot Start II DNA polymerase— extremely short PCR protocol times
  • Master Mix format with premixed gel loading dyebold — minimizes possibility of cross-contamination, reduces sample handling and allows direct loading on gel 
Specifications
Specifications
500
Direct PCR reagent
500 x 50μL rxns
-20°C