Learn More
Thermo Scientific™ Phire Tissue Direct PCR Master Mix Promotion
Amplify DNA directly from variety of human and animal samples with no prior purification with Phire Tissue Direct PCR Master Mix
Manufacturer:  Thermo Scientific™ F170L
 View more versions of this product
Catalog No. F170LPM
Add to cart
Description
Description
  • Direct PCR – sample is added directly to PCR reaction, therefore therese is no need for time-consuming and expensive DNA purification steps
  • Dilution & Storage protocol available – allows tens of PCR reactions from one tiny sample and allows re-testing
  • Specially engineered Phire DNA polymerase – extremely short PCR protocol times
  • Master Mix format with premixed gel loading dye – minimizes possibility of cross-contamination, reduces sample handling and allows direct loading on gel
Specifications
Specifications
Genotyping, Transgene detection, Knockout analysis, Sequencing
1250
Store all components of kit at -20°C upon arrival. The Dilution Buffer can also be stored at 4°C once it is thawed
2X Phire Tissue Direct PCR Master Mix, Dilution Buffer, DNARelease Additive, Universal control primer mix (25μM each), nuclease- free Water, O'GeneRuler Express DNA Ladder
1250 x 20μL Reactions
2X