Learn More
PHKG2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 151091AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-172-9986
Only null left
Add to Cart
Description
Description
Phosphorylase b kinase catalyzes the phosphorylation of serine in certain substrates, including troponin I. This isozyme may regulate glycogeneolysis in the testis.
Specifications
Specifications
PHKG2
Polyclonal
Unconjugated
PHKG2
GSD9C, PHK gamma T, PHKG2, PHK-gamma-LT, Phosphorylase kinase subunit gamma-2, PSK C3
Rabbit
Antigen Affinity Chromatography
RUO
140671, 5261, 68961
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P15735, P31325, Q9DB30
PHKG2
PHKG2 Fusion Protein Ag6829
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.