Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PHLDA1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$482.50
Specifications
Antigen | PHLDA1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP153066
|
Novus Biologicals
NBP153066 |
100 μL |
Each of 1 for $482.50
|
|
|||||
Description
PHLDA1 Polyclonal specifically detects PHLDA1 in Human samples. It is validated for Western Blot.Specifications
PHLDA1 | |
Polyclonal | |
Rabbit | |
Apoptosis-associated nuclear protein, DT1P1B11, MGC131738, PHRIPT-cell death-associated gene 51 protein, pleckstrin homology-like domain family A member 1, pleckstrin homology-like domain, family A, member 1, PQ-rich protein, Proline- and glutamine-rich protein, Proline- and histidine-rich protein, proline-histidine rich protein, TDAG51PQR protein | |
PHLDA1 | |
IgG | |
45 kDa |
Western Blot | |
Unconjugated | |
RUO | |
22822 | |
Synthetic peptides corresponding to PHLDA1(pleckstrin homology-like domain, family A, member 1) The peptide sequence was selected from the C terminal of PHLDA1. Peptide sequence LAVKSTRQKQQHLVQQQPPSQPQPQPQLQPQPQPQPQPQPQPQSQPQPQP. | |
Primary |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title