Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CENTiCARE Phlebotomy Laptop Carts

Designed with many storage options and deluxe 4 in. twin wheel casters

Manufacturer:  CENTiCARE CPHL1601H

 View more versions of this product

Catalog No. 22-555-152


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Drawer: Shallow w/Glove box mounted on the sside; Regular w/o Glove box mounted on the side; w/ Green frog, swing lid; Colorful stenciling on legs of base
Cart - 1601H
Drawer: Blue (Shallow); Red (Regular)
Provides mobile work station for phlebotomy, nursing units and IV start teams that need laptop and supplies handy
Hex keys and extra set screws for assembly
180 Days Limited Liability
8L
Shallow: O.D 43.1 x 38.1 x 12.7cm (17 x 15 x 5 in.); Regular: 43.1 x 38.1 x 20.3cm (17 x 15 x 8 in.)
Fisher 14-827-122 container
Large
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit