Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phlorizin, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
50mg
This item is not returnable. View return policy
60-81-1
C21H24O10
436.413
IOUVKUPGCMBWBT-QNDFHXLGSA-N
phlorizin,phloridzin,phlorhizin,phlorizoside,floridzin,phlorrhizin,phloretin 2'-glucoside,phloridzosid,phlorrhizen,phlorizine
6072
CHEBI:8113
1-[2,4-dihydroxy-6-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyphenyl]-3-(4-hydroxyphenyl)propan-1-one
C1=CC(=CC=C1CCC(=O)C2=C(C=C(C=C2OC3C(C(C(C(O3)CO)O)O)O)O)O)O
This item is not returnable. View return policy
60-81-1
C21H24O10
436.413
IOUVKUPGCMBWBT-QNDFHXLGSA-N
phlorizin,phloridzin,phlorhizin,phlorizoside,floridzin,phlorrhizin,phloretin 2'-glucoside,phloridzosid,phlorrhizen,phlorizine
6072
CHEBI:8113
1-[2,4-dihydroxy-6-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyphenyl]-3-(4-hydroxyphenyl)propan-1-one
C1=CC(=CC=C1CCC(=O)C2=C(C=C(C=C2OC3C(C(C(C(O3)CO)O)O)O)O)O)O

Na+-glucose cotransporter (SGLT) inhibitor

Inhibitor of Na+-glucose cotransporters (SGLT). Produces renal glycosuria and blocks intestinal glucose absorption. Normalizes insulin sensitivity in diabetic rats.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 50mg
Formula Weight 436.41
Percent Purity >99%
Chemical Name or Material Phlorizin

Recommended Storage : Store at -20°C