Learn More
Alfa Aesar™ Phloroglucide, 95%
Specifications
Sensitivity | Light sensitive |
Melting Point | ~296°C (decomposition) |
Quantity | 5g |
Beilstein | 3324960 |
Formula Weight | 234.21 |
Percent Purity | 95% |
Chemical Name or Material | Phloroglucide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
TSCA : No
Recommended Storage : Ambient temperatures