Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phloroglucinol Dihydrate, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
6099-90-7
C6H6O3·2H2O
162.14
MFCD00149090
MPYXTIHPALVENR-UHFFFAOYSA-N
phloroglucinol dihydrate,benzene-1,3,5-triol dihydrate,1,3,5-trihydroxybenzene dihydrate,1,3,5-benzenetriol, dihydrate,phloroglucinol, dihydrate,sym-trihydroxybenze,unii-0x7b22brg1,ccris 6521,1,3,5-benzoltrioldihydrat,phloroglucinoldihydrate
80196
benzene-1,3,5-triol;dihydrate
C1=C(C=C(C=C1O)O)O.O.O
6099-90-7
C6H6O3·2H2O
162.14
MFCD00149090
MPYXTIHPALVENR-UHFFFAOYSA-N
phloroglucinol dihydrate,benzene-1,3,5-triol dihydrate,1,3,5-trihydroxybenzene dihydrate,1,3,5-benzenetriol, dihydrate,phloroglucinol, dihydrate,sym-trihydroxybenze,unii-0x7b22brg1,ccris 6521,1,3,5-benzoltrioldihydrat,phloroglucinoldihydrate
80196
benzene-1,3,5-triol;dihydrate
C1=C(C=C(C=C1O)O)O.O.O
Melting Point 218.0°C to 221.0°C
Quantity 500g
Packaging Glass bottle
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Linear Formula C6H3(OH)3·2H2O
Beilstein 06, 1092
Merck Index 15, 7439
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and pyridine
Formula Weight 162.14
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material Phloroglucinol dihydrate
Show More Show Less

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning