Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phloroglucinol Reagent, Reagents
Phloroglucinol Reagent, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Reagents Holdings Llc CP10720020E7
Quantity | 20 L |
---|
Chemical Identifiers
67-56-1, 7732-18-5, 7647-01-0 | |
32.04 | |
1,3,5-Trihydroxybenzene Dihydrate, 1,3,5-Benzenetriol Dihydrate | |
CO |
CH4O | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol |
Specifications
Yellow | |
CH4O | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol | |
1.3% | |
HDPE Winpak | |
<1 | |
Non-sterile | |
Phloroglucinol Reagent |
67-56-1, 7732-18-5, 7647-01-0 | |
1,3,5-Trihydroxybenzene Dihydrate, 1,3,5-Benzenetriol Dihydrate | |
CO | |
32.04 | |
Reagent | |
0.98 | |
20 L | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction