Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phoenix Diagnostics Electrodes for Bayer™ Rapidlab™ Analyzers

Equivalent performance and reliability as Bayer-manufactured products.

Manufacturer:  Phoenix Diagnostics E100

 View more versions of this product

Catalog No. 23-293-166


  Check Availability
Add to cart

Specifications

Description

For use with Bayer Rapidlab Series Analyzers.

Order Info

Reference and pH fill solutions available separately (Cat. Nos. 23-293-174 and -175).

Description & Specifications

Specifications

pH Electrode
Bayer Rapidlab 200 Series, 348, 634, 800 Series
Blood gas analysis
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit