Phoenix Diagnostics Electrodes for Bayer™ Rapidlab™ Analyzers
Equivalent performance and reliability as Bayer-manufactured products.
Manufacturer: Phoenix Diagnostics E140
 View more versions of this product
Catalog No. 23-293-168
    N/A
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 
    Specifications
    Product Type Sodium Electrode
    For Use With (Equipment) Bayer Rapidlab 288, 348, 614, 644, 654, 664, 800 Series
    For Use With (Application) Blood gas analysis
    For use with Bayer Rapidlab Series Analyzers.
    Order Info
    Reference and pH fill solutions available separately (Cat. Nos. 23-293-174 and -175).
  • Description & Specifications
Related Products
Product Content Feedback