Learn More
Phoenix Diagnostics Electrodes for Bayer™ Rapidlab™ Analyzers
Equivalent performance and reliability as Bayer-manufactured products.
Manufacturer:  Phoenix Diagnostics E160
 View more versions of this product
Catalog No. 23-293-170
Due to product restrictions, please to purchase or view availability for this product.
Specifications
Description
For use with Bayer Rapidlab Series Analyzers.
Order Info
Reference and pH fill solutions available separately (Cat. Nos. 23-293-174 and -175).
Description & Specifications
Specifications
Chloride Electrode
Bayer Rapidlab 288, 348, 644, 850, 855, 860, 865
Blood gas analysis