Phoenix Diagnostics Electrodes for Bayer™ Rapidlab™ Analyzers
Equivalent performance and reliability as Bayer-manufactured products.
Manufacturer: Phoenix Diagnostics E170
 View more versions of this product
Catalog No. 23-293-171
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Product Type Calcium Electrode
    For Use With (Equipment) Bayer Rapidlab 288, 348, 634, 850, 855, 860, 865
    For Use With (Application) Blood gas analysis
    For use with Bayer Rapidlab Series Analyzers.
    Order Info
    Reference and pH fill solutions available separately (Cat. Nos. 23-293-174 and -175).
  • Description & Specifications
Related Products
Product Content Feedback