Phoenix Diagnostics General Chemistry Linearity
Advanced product line of controls with extensive analyte breadth
Manufacturer: Phoenix Diagnostics, IncSupplier Diversity Partner PH5020
 View more versions of this product
Catalog No. 23-293-300
  Check Availability
Add to Cart
    Specifications
    Product Type Multi-analyte linearity control
    Detectable Analytes Total Protein, Albumin, Uric Acid, Creatinine, Glucose, Blood Urea Nitrogen, Lactate, Phosphorous, Sodium, Potassium, Calcium, Chloride, Lithium, Iron, Magnesium, Cholesterol, Triglyceride and Carbon Dioxide
    For Use With (Application) In-vitro diagnostic use in verifying reportable ranges and determining linearity in automated, semi-automated and manual chemistry systems
    For Use With (Equipment) Chemistry analyzer
    Quantity 1mL
  • Description & Specifications
Related Products
Product Content Feedback