Learn More
Phoenix Diagnostics General Chemistry Linearity
Advanced product line of controls with extensive analyte breadth
Manufacturer:  Phoenix Diagnostics, IncSupplier Diversity Partner PH5050
 View more versions of this product
Catalog No. 23-293-305
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Digitoxin, Digoxin, Homocysteine, C-reactive Protein, Myoglobin, NT-proBNP, Troponin-I, Troponin-T and Creatine Kinase-MB
Chemistry analyzer
Cardiac marker linearity control
In-vitro diagnostic use in verifying reportable ranges and determining linearity in automated, semi-automated and manual chemistry systems
1mL