Phoenix Diagnostics General Chemistry Linearity
Advanced product line of controls with extensive analyte breadth
Manufacturer: Phoenix Diagnostics, IncSupplier Diversity Partner PH5060
 View more versions of this product
Catalog No. 23-293-306
  Check Availability
Add to Cart
    Specifications
    Product Type Ammonia/Ethanol linearity control
    Detectable Analytes Ammonia and Ethyl Alcohol
    For Use With (Application) In-vitro diagnostic use in verifying reportable ranges and determining linearity in automated, semi-automated and manual chemistry systems
    For Use With (Equipment) Chemistry analyzer
    Quantity 1mL
  • Description & Specifications
Related Products
Product Content Feedback