Phoenix Diagnostics Olympus™ Electrodes
Reference Electrodes for Olympus AU Series analyzers.
Manufacturer: Phoenix Diagnostics E930
 View more versions of this product
Catalog No. 22-293-264
  Check Availability
Add to Cart
    Specifications
    Product Type Na+ Electrode
    For Use With Olympus™ AU series analyzer
    • Replacement electrodes available for use in all Olympus AU series analyzers
    • Na+, K+, Cl-, and Reference electrodes available
  • Description & Specifications
Related Products
Product Content Feedback