Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phoenix Diagnostics Supplies for Bayer™ Rapidlab™ 800 Series Analyzers

For use with Bayer Rapidlab 800 Series Analyzers.

Manufacturer:  Phoenix Diagnostics 03801

 View more versions of this product

Catalog No. 23-293-161


  Check Availability
Add to cart

Specifications

Description

  • For Bayer Rapidlab Models 840, 845, 850, 855, 860, 865 Analyzers (except Cat. No. 23-293-165: 865 only)
Description & Specifications

Specifications

Buffer
Bayer Rapidlab Models 840, 845, 850, 855, 860, 865 Analyzers (except Cat. No. 23-293-165: 865 only)
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit