Learn More
Phoenix Diagnostics Supplies for Bayer™ Rapidlab™ 800 Series Analyzers
For use with Bayer Rapidlab 800 Series Analyzers.
Manufacturer:  Phoenix Diagnostics 03802
 View more versions of this product
Catalog No. 23-293-162
Due to product restrictions, please to purchase or view availability for this product.
Specifications
Description
  • For Bayer Rapidlab Models 840, 845, 850, 855, 860, 865 Analyzers (except Cat. No. 23-293-165: 865 only)
Description & Specifications
Specifications
Wash Zero Reagent
Bayer Rapidlab Models 840, 845, 850, 855, 860, 865 Analyzers (except Cat. No. 23-293-165: 865 only)