Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phorbol 12-myristate 13-acetate, 97%, ACROS Organics™

Click to view available options
Quantity:
1mg
5mg
Packaging:
Glass bottle
16561-29-8
C36H56O8
616.84
MFCD00036736
PHEDXBVPIONUQT-RGYGYFBISA-N
phorbol 12-myristate 13-acetate,phorbol ester,12-o-tetradecanoylphorbol-13-acetate,12-o-tetradecanoylphorbol 13-acetate,factor a1,tetradecanoylphorbol acetate,12-tetradecanoylphorbol 13-acetate,factor a1 croton oil,phorbol 13-acetate 12-myristate,phorbol-12-myristate-13-acetate
27924
CHEBI:37537
CCCCCCCCCCCCCC(=O)OC1C(C2(C(C=C(CC3(C2C=C(C3=O)C)O)CO)C4C1(C4(C)C)OC(=O)C)O)C
16561-29-8
C36H56O8
616.84
MFCD00036736
PHEDXBVPIONUQT-RGYGYFBISA-N
phorbol 12-myristate 13-acetate,phorbol ester,12-o-tetradecanoylphorbol-13-acetate,12-o-tetradecanoylphorbol 13-acetate,factor a1,tetradecanoylphorbol acetate,12-tetradecanoylphorbol 13-acetate,factor a1 croton oil,phorbol 13-acetate 12-myristate,phorbol-12-myristate-13-acetate
27924
CHEBI:37537
CCCCCCCCCCCCCC(=O)OC1C(C2(C(C=C(CC3(C2C=C(C3=O)C)O)CO)C4C1(C4(C)C)OC(=O)C)O)C
Color White to Yellow
Quantity 1mg
Melting Point 72.0°C
Packaging Glass bottle
Assay Percent Range 96% min. (HPLC)
Merck Index 14,7332
Formula Weight 616.84
Physical Form Fine Crystalline Powder
Percent Purity 97%
Chemical Name or Material Phorbol 12-myristate 13-acetate

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning