Learn More
phos-AMPKα (Thr172) Rabbit anti-Human,Mouse, KHL, Clone: 5J23, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma ZRB12134X25UL
 View more versions of this product
Catalog No. ZRB12134XMI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
phos-AMPKα (Thr172) Monoclonal antibody specifically detects phos-AMPKα (Thr172) in Human,Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry, Peptide Inhibition Assay, Western Blotting.
Specifications
Specifications
phos-AMPKα (Thr172)
5J23
KHL
Purified
PRKAA2, AMPK, AMPK2
KLH-conjugated linear peptide corresponding to 10 amino acids surrounding phosphothreonine 172 from human AMPK alpha 2.
Target Molecular Weight 62.32 kDa calculated; ∼62 kDa observed. Uncharacterized bands may be observed in some lysate(s).
4 x 25 μL
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Monoclonal
Human, Mouse
Immunocytochemistry, Immunohistochemistry, Inhibition Assays, Western Blot
300 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
NP_006243.2, NP_006242.5
5'-AMP-activated protein kinase catalytic subunit alpha-2;EC: 2.7.11.1;AMPK subunit alpha-2;Acetyl-CoA carboxylase kinase;ACACA kinase;Hydroxymethylglutaryl-CoA reductase kinase;HMGCR kinase
Rabbit
IgG
Protein A Purified
Signaling
Primary
4X