Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosgene iminium chloride, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
33842-02-3
C3H6Cl3N
162.45
MFCD00011870
NRNFKRFWZQQDMD-UHFFFAOYSA-M
dichloromethylene dimethylammonium chloride,phosgeniminium chloride,n-dichloromethylene-n-methylmethanaminium chloride,methanaminium, n-dichloromethylene-n-methyl-, chloride,dichloromethylene-dimethyliminium chloride,dichloromethylenedimethylammonium chloride,dichloromethylidene dimethylazanium chloride,phosgeneiminium chloride,phosgene-iminium chloride
2723945
dichloromethylidene(dimethyl)azanium;chloride
C[N+](=C(Cl)Cl)C.[Cl-]
33842-02-3
C3H6Cl3N
162.45
MFCD00011870
NRNFKRFWZQQDMD-UHFFFAOYSA-M
dichloromethylene dimethylammonium chloride,phosgeniminium chloride,n-dichloromethylene-n-methylmethanaminium chloride,methanaminium, n-dichloromethylene-n-methyl-, chloride,dichloromethylene-dimethyliminium chloride,dichloromethylenedimethylammonium chloride,dichloromethylidene dimethylazanium chloride,phosgeneiminium chloride,phosgene-iminium chloride
2723945
dichloromethylidene(dimethyl)azanium;chloride
C[N+](=C(Cl)Cl)C.[Cl-]
Melting Point 181°C to 185°C
Infrared Spectrum Authentic
Packaging Glass bottle
Color Yellow
Quantity 5g
Assay Percent Range 97.5% min. (Argentometry)
Linear Formula Cl2C=N(CH3)2Cl
Fieser 04,135; 05,195; 06,170; 08,156
Merck Index 15, 10174
Solubility Information Solubility in water: hydrolysis
Formula Weight 162.45
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material Phosgene iminium chloride, 98%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 251-695-4