Learn More
Aat BioquestSupplier Diversity Partner PhosLite™ Green  DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 11630
This water-soluble PhosLite™ Green substrate generates a bright and photostable yellow-green fluorescent precipitate at the site of phosphtase activity.
Catalog No. 50-195-3374