Learn More
Supply SolutionsSupplier Diversity Partner Hach Company Phosphorus (Reactive) Tnt Reagent Set. Parameter: Phosphate, Ortho; Method Name: Molybdovanadate; Range: 1.0 To 100.0 Mg/L Po4 DFS Item
Manufacturer:  Supply SolutionsSupplier Diversity Partner HACH 2767345
For determination of Reactive Phosphorus (ortho-phosphate) by the Molybdovanadate Test 'N Tube Method. Hach Method 8114. Range: 1.0 - 100.0 mgL PO4. Set includes vials and reagents for up to 50 tests (dependent upon the number of reagent blank tests performed).; Parameter: Phosphate, ortho; Method Name: Molybdovanadate; Range: 1.0 to 100.0 mg/L PO4; Method: 8114; Platform : Test N Tube; Test 'N Tube products provide safe, convenient testing; Easy ordering
Catalog No. 50-476-293