Learn More
Alfa Aesar™ Phosphatase buffer 1 (5X)
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J62267AE
 View more versions of this product
Catalog No. AAJ62267AE
Order in Bulk
Request bulk or custom formats
Add to cart
Description
Description
  • Formulation: 100mM Tris (pH 7.5) and 100mM magnesium chloride
Specifications
Specifications
Phosphatase buffer 1 (5X)
Liquid
100mM Tris (pH 7.5) and 100mM magnesium chloride
100mL