Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphate, 0.5M buffer soln., pH 7.0, Alfa Aesar™

Click to view available options
Quantity:
250mL
500mL
7558-80-7
H2NaO4P
119.976
MFCD00003527
AJPJDKMHJJGVTQ-UHFFFAOYSA-M
23672064
CHEBI:37585
sodium;dihydrogen phosphate
OP(=O)(O)[O-].[Na+]
7558-80-7
H2NaO4P
119.976
MFCD00003527
AJPJDKMHJJGVTQ-UHFFFAOYSA-M
23672064
CHEBI:37585
sodium;dihydrogen phosphate
OP(=O)(O)[O-].[Na+]
Concentration 0.5M buffer soln.
Physical Form Liquid
Packaging Plastic bottle
Quantity 250mL
Chemical Name or Material Phosphate

TSCA : Yes

Recommended Storage : Ambient temperatures