Learn More
Thermo Scientific Chemicals Phosphate, 0.5M buffer soln., pH 7.4
Description
Phosphate-buffered saline (abbreviated PBS) is a buffer solution commonly used in biological research. A high buffering capacity reagent used in molecular biology, biochemistry, and chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Concentration | 0.5M buffer soln. |
Molecular Weight (g/mol) | 119.976 |
Quantity | 250 mL |
Packaging | Plastic Bottle |
Content And Storage | Ambient Temperatures |
Form | Liquid |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.